accessibility menu, dialog, popup

UserName

CH3H2NH3CNH2HClHCl

CAS RN 76787-89-8

IUPAC Name: 3,3'-diethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride
Synonyms: 3,3'-diethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride
Molecular Weight (g/mol): 313.27
Molecular Formula: C16H22Cl2N2
InChi Key: DQPPOEIMMLEMPW-UHFFFAOYSA-N
SMILES: Cl.Cl.CCC1=C(N)C=CC(=C1)C1=CC(CC)=C(N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results