missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dibromo-5,5'-bis(trimethylsilyl)-2,2'-bithiophene 98.0+%, TCI America™

Catalog No. p-7128009
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39411G 1 g
D39415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39411G Supplier TCI America Supplier No. D39411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 207742-50-5
Molecular Formula C14H20Br2S2Si2
Molecular Weight (g/mol) 468.412
InChI Key ZKCVPMCCGPMMBH-UHFFFAOYSA-N
PubChem CID 15860540
IUPAC Name [4-bromo-5-(3-bromo-5-trimethylsilylthiophen-2-yl)thiophen-2-yl]-trimethylsilane
SMILES C[Si](C)(C)C1=CC(=C(S1)C2=C(C=C(S2)[Si](C)(C)C)Br)Br

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 1 g
Formula Weight 468.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dibromo-5,5′-bis(trimethylsilyl)-2,2′-bithiophene