accessibility menu, dialog, popup

UserName

CAS RN 207742-50-5

IUPAC Name: [3,3'-dibromo-5'-(trimethylsilyl)-[2,2'-bithiophen]-5-yl]trimethylsilane
Synonyms: [3,3'-dibromo-5'-(trimethylsilyl)-[2,2'-bithiophen]-5-yl]trimethylsilane
Molecular Weight (g/mol): 468.41
Molecular Formula: C14H20Br2S2Si2
InChi Key: ZKCVPMCCGPMMBH-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=CC(Br)=C(S1)C1=C(Br)C=C(S1)[Si](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results