missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dibromo-2,2'-bithiophene 98.0+%, TCI America™

$69.31 - $764.30

Chemical Identifiers

CAS 51751-44-1
Molecular Formula C8H4Br2S2
Molecular Weight (g/mol) 324.05
MDL Number MFCD00114806
InChI Key KBRZCEVRNLKHAZ-UHFFFAOYSA-N
PubChem CID 819087
IUPAC Name 3,3'-dibromo-2,2'-bithiophene
SMILES BrC1=C(SC=C1)C1=C(Br)C=CS1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D37981G
View Documents
TCI America
D37981G
1 g
Each for $69.31
Only null left
 
D37985G
View Documents
TCI America
D37985G
5 g
Each for $223.33
Only null left
 
D379825G
View Documents
TCI America
D379825G
25 g
Each for $764.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

51751-44-1
324.05
KBRZCEVRNLKHAZ-UHFFFAOYSA-N
3,3'-dibromo-2,2'-bithiophene
C8H4Br2S2
MFCD00114806
819087
BrC1=C(SC=C1)C1=C(Br)C=CS1

Specifications

51751-44-1
White-Yellow
MFCD00114806
KBRZCEVRNLKHAZ-UHFFFAOYSA-N
3,3'-dibromo-2,2'-bithiophene
819087
≥98.0% (GC)
3,3′-Dibromo-2,2′-bithiophene
99°C
C8H4Br2S2
1 g
BrC1=C(SC=C1)C1=C(Br)C=CS1
324.05
324.05
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents