missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dibromo-2,2'-bithiophene 98.0+%, TCI America™

Catalog No. d37985g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
D37985G 5 g
D37981G 1 g
D379825G 25 g
3 options

Catalog No. D37985G

Supplier: TCI America D37985G

Only null left

Chemical Identifiers

CAS 51751-44-1
Molecular Formula C8H4Br2S2
Molecular Weight (g/mol) 324.05
MDL Number MFCD00114806
InChI Key KBRZCEVRNLKHAZ-UHFFFAOYSA-N
PubChem CID 819087
IUPAC Name 3,3'-dibromo-2,2'-bithiophene
SMILES BrC1=C(SC=C1)C1=C(Br)C=CS1

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 5 g
Formula Weight 324.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dibromo-2,2′-bithiophene
TSCA No
Recommended Storage Refrigerator