missing translation for 'onlineSavingsMsg'
Learn More

3,3″-Dibromo-1,1':3',1″-terphenyl 97.0+%, TCI America™

Catalog No. p-7128597
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4999200MG 200 mg
D49991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4999200MG Supplier TCI America Supplier No. D4999200MG
Only null left

Chemical Identifiers

CAS 95962-62-2
Molecular Formula C18H12Br2
Molecular Weight (g/mol) 388.102
MDL Number MFCD28138089
InChI Key FGKHIPSLESGJNR-UHFFFAOYSA-N
Synonym 3,3′C′C-Dibromo-m-terphenyl
PubChem CID 12088545
IUPAC Name 1,3-bis(3-bromophenyl)benzene
SMILES C1=CC(=CC(=C1)C2=CC(=CC=C2)Br)C3=CC(=CC=C3)Br

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 225°C
Quantity 200 mg
UN Number 3152
Formula Weight 388.10
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′′-Dibromo-1,1′:3′,1′′-terphenyl