missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzidine Tetrahydrochloride Hydrate 98.0+%, TCI America™

Catalog No. D37575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37575G 5 g
D37571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37575G Supplier TCI America Supplier No. D37575G

Chemical Identifiers

CAS 868272-85-9
Molecular Formula C12H18Cl4N4
Molecular Weight (g/mol) 360.10
MDL Number MFCD08273058
InChI Key KJDSORYAHBAGPP-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzidine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, acmc-209qbm, 3,3'-diaminobenzidine hydrate tetrahydrochloride, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se
PubChem CID 23191111
IUPAC Name [1,1'-biphenyl]-3,3',4,4'-tetramine tetrahydrochloride
SMILES Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1

Specifications

Melting Point 280°C
Color Purple-White
Quantity 5 g
Formula Weight 360.10
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diaminobenzidine Tetrahydrochloride Hydrate [for Biochemical Research]
RTECSNumber DV8753000
TSCA No
Recommended Storage Refrigerator