accessibility menu, dialog, popup

UserName

CAS RN 868272-85-9

IUPAC Name: [1,1'-biphenyl]-3,3',4,4'-tetramine tetrahydrochloride
Synonyms: 3,3'-diaminobenzidine tetrahydrochloride
Molecular Weight (g/mol): 360.1
Molecular Formula: C12H18Cl4N4
InChi Key: KJDSORYAHBAGPP-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)