missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%

Catalog No. AAJ6221606
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6221606 5 g
AAJ6221609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAJ6221606 Supplier Thermo Scientific Chemicals Supplier No. J6221606
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

3,3'-Diaminobenzidine tetrahydrochloride hydrate, CAS # 868272-85-9, is a peroxidase substrate that is commonly used for immunoblotting and immunohistochemical staining techniques. | CAS: 868272-85-9 | C12H18Cl4N4 | 360.10 g/mol

3,3?-diaminobenzidine-tetrahydrochloride-dihydrate (DAB) was used as a chromogen, for detecting the antigen-antibody complex. It is one of the most potent marker being used in immunochemistry, mostly at the electron and light microscope levels. It is used as a histological stain for peroxidase activity. Reaction between 3,3'-Diaminobenzidine and peroxidase forms a visible insoluble product. This approach has been used to assay activity of glutamate decarboxylase in rat cerebellar sections and to detect horseradish peroxidase presence on cultured cell surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 3,3'-Diaminobenzidine tetrahydrochloride hydrate is an organic compound used as a peroxidase substrate

Application

• 3,3'-Diaminobenzidine tetrahydrochloride hydrate is commonly used for immunoblotting and immunohistochemical staining techniques

• It has also been used for the histological stain to measure the peroxidase activity. The interaction between 3,3'-Diaminobenzidine tetrahydrochloride hydrate and peroxidase leads to the formation of a visible insoluble product. Therefore, it has been applied to measure glutamate decarboxylase activity in rat cerebellar sections and to detect horseradish peroxidase presence on cultured cell surfaces

• It is used as a chromogen for detecting the antigen-antibody complex

• This compound is also used in several laboratory applications, including electron and light microscope, and FISH (fluorescence in situ hybridization)

Chemical Identifiers

CAS 868272-85-9
Molecular Formula C12H18Cl4N4
Molecular Weight (g/mol) 360.10
MDL Number MFCD08273058
InChI Key KJDSORYAHBAGPP-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzidine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, acmc-209qbm, 3,3'-diaminobenzidine hydrate tetrahydrochloride, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se
PubChem CID 23191111
IUPAC Name 4-(3,4-diaminophenyl)benzene-1,2-diamine;hydrate
SMILES Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1

Specifications

Color Gray
Quantity 5 g
Solubility Information Soluble at 5%(w/v) in water
Formula Weight 360.11 (Anhydrous)
Physical Form Powder
Chemical Name or Material 3,3'-Diaminobenzidine tetrahydrochloride hydrate
Hazard Category H315-H320-H335-H341-H350
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only