missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Di-tert-butyl-5,5'-dimethyldiphenoquinone 97.0+%, TCI America™

Catalog No. D44071G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D44071G Supplier TCI America Supplier No. D44071G
Only null left

Chemical Identifiers

CAS 2417-00-7
Molecular Formula C22H28O2
Molecular Weight (g/mol) 324.464
MDL Number MFCD01310671
InChI Key UTSXCYFVGSFPGR-FOCLMDBBSA-N
PubChem CID 5355774
IUPAC Name (4E)-2-tert-butyl-4-(3-tert-butyl-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-6-methylcyclohexa-2,5-dien-1-one
SMILES CC1=CC(=C2C=C(C(=O)C(=C2)C(C)(C)C)C)C=C(C1=O)C(C)(C)C

Specifications

Melting Point 200°C
Color Purple-Red
Quantity 1 g
Formula Weight 324.46
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Di-tert-butyl-5,5′-dimethyldiphenoquinone