accessibility menu, dialog, popup

UserName

CAS RN 2417-00-7

CH3OH3CH3CCH3CH3OCH3H3CCH3(E)

CAS RN 2417-00-7

IUPAC Name: (E)-3,3'-di-tert-butyl-5,5'-dimethyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione
Synonyms: (E)-3,3'-di-tert-butyl-5,5'-dimethyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione
Molecular Weight (g/mol): 324.46
Molecular Formula: C22H28O2
InChi Key: UTSXCYFVGSFPGR-FOCLMDBBSA-N
SMILES: CC1=C\C(C=C(C1=O)C(C)(C)C)=C1\C=C(C)C(=O)C(=C1)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results