missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenyl)propionic acid, 97%

Catalog No. AAL1808006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1808006 5 g
AAL1808003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1808006 Supplier Thermo Scientific Chemicals Supplier No. L1808006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42287-90-1 | C9H9BrO2 | 229.073 g/mol

It is used in the synthesis and characterization of monoisomeric 1,8,15,22-substituted (A3B and A2B2) phthalocyanines and phthalocyanine-fullerene dyads.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and characterization of monoisomeric 1,8,15,22-substituted (A3B and A2B2) phthalocyanines and phthalocyanine-fullerene dyads.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 42287-90-1
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD01310792
InChI Key DWKWMFSWLCIMKI-UHFFFAOYSA-N
Synonym 3-3-bromophenyl propanoic acid, 3-3-bromophenyl propionic acid, 3-3-bromo-phenyl-propionic acid, 3-bromophenylpropanoic acid, acmc-209jor, m-bromohydrocinnamic acid, 3-bromohydrocinnamic acid, benzenepropanoic acid,3-bromo, 3-3-bromophenyl propionicacid, 3-m-bromophenyl propionic acid
PubChem CID 2063862
IUPAC Name 3-(3-bromophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)Br)CCC(=O)O

Specifications

Melting Point 63°C to 65°C
Quantity 5 g
Beilstein 2362753
Solubility Information Insoluble in water.
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material 3-(3-Bromophenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only