accessibility menu, dialog, popup

UserName

CAS RN 42287-90-1

IUPAC Name: 3-(3-bromophenyl)propanoic acid
Synonyms: 3-(3-bromophenyl)propanoic acid
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: DWKWMFSWLCIMKI-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results