missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenyl)-5-methyl-1,2,4-oxadiazole, ≥97%, Thermo Scientific™

Catalog No. CC39110CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC39110CB 250mg
CC39110DA 1g
CC39110DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC39110CB Supplier Thermo Scientific Chemicals Supplier No. CC39110CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 160377-57-1
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
MDL Number MFCD08087563
InChI Key GTYLSVIVKRJHIQ-UHFFFAOYSA-N
Synonym 3-3-bromophenyl-5-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-3-bromophenyl-5-methyl, acmc-1c5gb, chembrdg-bb 9035421, 3-3-bromophenyl-5-methyl-1,2,4-oxadiazol, 3-3-bromophenyl-5-methyl-1,2,4 oxadiazole, 3-3-bromophenyl-5-methyl-1,2,4-oxadiazole saltdata: free
PubChem CID 8895139
IUPAC Name 3-(3-bromophenyl)-5-methyl-1,2,4-oxadiazole
SMILES CC1=NC(=NO1)C2=CC(=CC=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 3-(3-Bromophenyl)-5-methyl-1, 2, 4-oxadiazole
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.