CAS RN 160377-57-1
CAS RN 160377-57-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(3-Bromophenyl)-5-methyl-1,2,4-oxadiazole, ≥97%, Thermo Scientific™
CAS: 160377-57-1 Molecular Formula: C9H7BrN2O Molecular Weight (g/mol): 239.072 MDL Number: MFCD08087563 InChI Key: GTYLSVIVKRJHIQ-UHFFFAOYSA-N Synonym: 3-3-bromophenyl-5-methyl-1,2,4-oxadiazole,1,2,4-oxadiazole, 3-3-bromophenyl-5-methyl,acmc-1c5gb,chembrdg-bb 9035421,3-3-bromophenyl-5-methyl-1,2,4-oxadiazol,3-3-bromophenyl-5-methyl-1,2,4 oxadiazole,3-3-bromophenyl-5-methyl-1,2,4-oxadiazole saltdata: free PubChem CID: 8895139 IUPAC Name: 3-(3-bromophenyl)-5-methyl-1,2,4-oxadiazole SMILES: CC1=NC(=NO1)C2=CC(=CC=C2)Br