missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenyl)-1H-pyrazole, 98%, Thermo Scientific Chemicals

Catalog No. AAH5043806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5043806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5043806 Supplier Thermo Scientific Chemicals Supplier No. H5043806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149739-65-1 | Molecular Formula: C9H7BrN2 | Molecular Weight: 223.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 149739-65-1
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.07
MDL Number MFCD01940433
InChI Key NVRXIZHZQPRBKL-UHFFFAOYSA-N
Synonym 3-3-bromophenyl-1h-pyrazole, 5-3-bromophenyl-1h-pyrazole, 5-3-bromo-phenyl-1h-pyrazole, 3-3-bromophenyl pyrazole, 3-3-bromophenyl-2h-pyrazole, 1h-pyrazole, 3-3-bromophenyl, 1-bromo-3-1h-pyrazol-3-yl benzene, 5-3-bromophenyl pyrazole, acmc-1c2w8, ksc494i0r
PubChem CID 2735613
IUPAC Name 5-(3-bromophenyl)-1H-pyrazole
SMILES BrC1=CC=CC(=C1)C1=CC=NN1

Specifications

Quantity 5 g
Formula Weight 223.07
Percent Purity 98%
Chemical Name or Material 3-(3-Bromophenyl)-1H-pyrazole
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only