missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Bithiophene 98.0+%, TCI America™

Catalog No. B39061G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B39061G Supplier TCI America Supplier No. B39061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3172-56-3
Molecular Formula C8H6S2
Molecular Weight (g/mol) 166.256
MDL Number MFCD00041214
InChI Key IAAQEGBHNXAHBF-UHFFFAOYSA-N
Synonym 3,3′C-Bithienyl
PubChem CID 76638
IUPAC Name 3-thiophen-3-ylthiophene
SMILES C1=CSC=C1C2=CSC=C2

Specifications

Melting Point 133°C
Boiling Point 180°C
Quantity 1 g
Formula Weight 166.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Bithiophene