accessibility menu, dialog, popup

UserName

CAS RN 3172-56-3

IUPAC Name: 3,3'-bithiophene
Synonyms: 3,3'-bithiophene
Molecular Weight (g/mol): 166.26
Molecular Formula: C8H6S2
InChi Key: IAAQEGBHNXAHBF-UHFFFAOYSA-N
SMILES: S1C=CC(=C1)C1=CSC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results