missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Bis(trifluoromethyl)benzophenone 98.0+%, TCI America™

Catalog No. B32175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B32175G Supplier TCI America Supplier No. B32175G
Only null left

Chemical Identifiers

CAS 1868-00-4
Molecular Formula C15H8F6O
Molecular Weight (g/mol) 318.218
MDL Number MFCD00000390
InChI Key POOXOHISLGOAEF-UHFFFAOYSA-N
Synonym 3,3'-bis trifluoromethyl benzophenone, bis 3-trifluoromethyl phenyl methanone, 3,3'-di trifluoromethyl benzophenone, 3,3'-bis trifluoromethyl ,benzophenone, di3-trifluoromethyl phenyl ketone, pubchem6402, acmc-1chtz, 3,3-bis trifluoromethyl benzophenone, labotest-bb lt00159871
PubChem CID 599788
IUPAC Name bis[3-(trifluoromethyl)phenyl]methanone
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)C2=CC(=CC=C2)C(F)(F)F

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
Formula Weight 318.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Bis(trifluoromethyl)benzophenone