missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Bis(bromomethyl)oxetane 95.0+%, TCI America™
$210.63 - $633.78
Chemical Identifiers
| CAS | 2402-83-7 |
|---|---|
| Molecular Formula | C5H8Br2O |
| Molecular Weight (g/mol) | 243.93 |
| MDL Number | MFCD00269694 |
| InChI Key | QOPMHMFIIMJWET-UHFFFAOYSA-N |
| Synonym | 2,2-Bis(bromomethyl)-1,3-epoxypropane |
| PubChem CID | 75464 |
| IUPAC Name | 3,3-bis(bromomethyl)oxetane |
| SMILES | BrCC1(CBr)COC1 |
Chemical Identifiers
| 2402-83-7 | |
| 243.93 | |
| QOPMHMFIIMJWET-UHFFFAOYSA-N | |
| 75464 | |
| BrCC1(CBr)COC1 |
| C5H8Br2O | |
| MFCD00269694 | |
| 2,2-Bis(bromomethyl)-1,3-epoxypropane | |
| 3,3-bis(bromomethyl)oxetane |
Specifications
| 2402-83-7 | |
| White-Yellow | |
| C5H8Br2O | |
| 1 g | |
| QOPMHMFIIMJWET-UHFFFAOYSA-N | |
| 3,3-bis(bromomethyl)oxetane | |
| 75464 | |
| ≥95.0% (GC) | |
| 3,3-Bis(bromomethyl)oxetane |
| 25°C | |
| 125°C | |
| MFCD00269694 | |
| 2,2-Bis(bromomethyl)-1,3-epoxypropane | |
| BrCC1(CBr)COC1 | |
| 243.93 | |
| 243.93 | |
| Crystalline Lumps |
Safety and Handling
TSCA : Yes