accessibility menu, dialog, popup

UserName

CAS RN 2402-83-7

BrBrO

CAS RN 2402-83-7

IUPAC Name: 3,3-bis(bromomethyl)oxetane
Synonyms: 3,3-bis(bromomethyl)oxetane
Molecular Weight (g/mol): 243.93
Molecular Formula: C5H8Br2O
InChi Key: QOPMHMFIIMJWET-UHFFFAOYSA-N
SMILES: BrCC1(CBr)COC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results