missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Bipyridyl 98.0+%, TCI America™
$298.41 - $1010.72
Chemical Identifiers
| CAS | 581-46-4 |
|---|---|
| Molecular Formula | C10H8N2 |
| Molecular Weight (g/mol) | 156.188 |
| MDL Number | MFCD00006371 |
| InChI Key | OFDVABAUFQJWEZ-UHFFFAOYSA-N |
| PubChem CID | 68487 |
| IUPAC Name | 3-pyridin-3-ylpyridine |
| SMILES | C1=CC(=CN=C1)C2=CN=CC=C2 |
Chemical Identifiers
| 581-46-4 | |
| 156.188 | |
| OFDVABAUFQJWEZ-UHFFFAOYSA-N | |
| 3-pyridin-3-ylpyridine |
| C10H8N2 | |
| MFCD00006371 | |
| 68487 | |
| C1=CC(=CN=C1)C2=CN=CC=C2 |
Specifications
| 581-46-4 | |
| 175°C | |
| MFCD00006371 | |
| 3,3′C-Bipyridine, 3,3′C-Dipyridyl | |
| C1=CC(=CN=C1)C2=CN=CC=C2 | |
| 156.188 | |
| 156.19 | |
| Liquid |
| Yellow | |
| C10H8N2 | |
| 1 g | |
| OFDVABAUFQJWEZ-UHFFFAOYSA-N | |
| 3-pyridin-3-ylpyridine | |
| 68487 | |
| ≥98.0% (GC,T) | |
| 3,3′-Bipyridyl |
Safety and Handling
RTECSNumber : DW1758000
TSCA : No