missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Bipyridyl 98.0+%, TCI America™

Catalog No. b39841g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B39841G 1 g
B39845G 5 g
2 options

Catalog No. B39841G

Supplier: TCI America B39841G

Only null left

Chemical Identifiers

CAS 581-46-4
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00006371
InChI Key OFDVABAUFQJWEZ-UHFFFAOYSA-N
PubChem CID 68487
IUPAC Name 3-pyridin-3-ylpyridine
SMILES C1=CC(=CN=C1)C2=CN=CC=C2

Specifications

Color Yellow
Boiling Point 175°C
Quantity 1 g
Synonym 3,3′C-Bipyridine, 3,3′C-Dipyridyl
Formula Weight 156.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,3′-Bipyridyl
RTECSNumber DW1758000
TSCA No