missing translation for 'onlineSavingsMsg'
Learn More

3,3',5-Triiodo-L-thyronine sodium salt

Catalog No. aaj63312me
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63312ME 500 mg
AAJ6331203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63312ME Supplier Thermo Scientific Chemicals Supplier No. J63312ME

CAS: 55-06-1 | C15H11I3NNaO4 | 672.959 g/mol

L-3,3',5-Triiodothyronine, Sodium Salt is shown to stimulate respiratory activity at the mitochondrial level. T3 is a thyroid hormone that increases rates of proteins synthesis, stimulates the breakdown of cholesterol, and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-3,3′,5-Triiodothyronine, Sodium Salt is shown to stimulate respiratory activity at the mitochondrial level. T3 is a thyroid hormone that increases rates of proteins synthesis, stimulates the breakdown of cholesterol, and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression.

Solubility
Partly miscible in water. Soluble in dimethyl sulfoxide, methanol, sodium hydroxide and ethanol. Practically insoluble in most other organic solvents.

Notes
Store at -20°C. Incompatible with oxidizing agents and heat. Light sensitive.

Chemical Identifiers

CAS 55-06-1
Molecular Formula C15H11I3NNaO4
Molecular Weight (g/mol) 672.959
MDL Number MFCD00002594
InChI Key SBXXSUDPJJJJLC-YDALLXLXSA-M
Synonym liothyronine sodium, cytomel, triostat, tertroxin, 3,3',5-triiodo-l-thyronine sodium salt, basoprocin, ibiothyron, cynomel, cytomel sodium, triiodothyronine sodium
PubChem CID 23666110
ChEBI CHEBI:6484
IUPAC Name sodium;(2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate
SMILES C1=CC(=C(C=C1OC2=C(C=C(C=C2I)CC(C(=O)[O-])N)I)I)O.[Na+]

Specifications

Melting Point 205°C
Color Brown to White
Quantity 500 mg
Beilstein 8179867
Sensitivity Light sensitive
Merck Index 14,5510
Solubility Information Partly miscible in water. Soluble in dimethyl sulfoxide,methanol,sodium hydroxide and ethanol. Practically insoluble in most other organic solvents.
Formula Weight 672.96
Physical Form Powder
Chemical Name or Material 3,3',5-Triiodo-L-thyronine sodium salt
EINECSNumber 200-223-5
RTECSNumber YP2836500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only