Learn More
3,3',5-Triiodo-L-thyronine sodium salt
CAS: 55-06-1 | C15H11I3NNaO4 | 672.959 g/mol
Supplier: Thermo Scientific Chemicals J6331203
Description
L-3,3',5-Triiodothyronine, Sodium Salt is shown to stimulate respiratory activity at the mitochondrial level. T3 is a thyroid hormone that increases rates of proteins synthesis, stimulates the breakdown of cholesterol, and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-3,3′,5-Triiodothyronine, Sodium Salt is shown to stimulate respiratory activity at the mitochondrial level. T3 is a thyroid hormone that increases rates of proteins synthesis, stimulates the breakdown of cholesterol, and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression.
Solubility
Partly miscible in water. Soluble in dimethyl sulfoxide, methanol, sodium hydroxide and ethanol. Practically insoluble in most other organic solvents.
Notes
Store at -20°C. Incompatible with oxidizing agents and heat. Light sensitive.
Specifications
| 3,3',5-Triiodo-L-thyronine sodium salt | |
| 205°C | |
| C15H11I3NNaO4 | |
| 1 g | |
| Light sensitive | |
| liothyronine sodium, cytomel, triostat, tertroxin, 3,3',5-triiodo-l-thyronine sodium salt, basoprocin, ibiothyron, cynomel, cytomel sodium, triiodothyronine sodium | |
| SBXXSUDPJJJJLC-YDALLXLXSA-M | |
| sodium;(2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate | |
| 23666110 | |
| 672.96 |
| 55-06-1 | |
| Brown to White | |
| MFCD00002594 | |
| 8179867 | |
| 14,5510 | |
| Partly miscible in water. Soluble in dimethyl sulfoxide,methanol,sodium hydroxide and ethanol. Practically insoluble in most other organic solvents. | |
| C1=CC(=C(C=C1OC2=C(C=C(C=C2I)CC(C(=O)[O-])N)I)I)O.[Na+] | |
| 672.959 | |
| CHEBI:6484 | |
| Powder |
Chemical Identifiers
| 55-06-1 | |
| 672.959 | |
| SBXXSUDPJJJJLC-YDALLXLXSA-M | |
| 23666110 | |
| sodium;(2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate |
| C15H11I3NNaO4 | |
| MFCD00002594 | |
| liothyronine sodium, cytomel, triostat, tertroxin, 3,3',5-triiodo-l-thyronine sodium salt, basoprocin, ibiothyron, cynomel, cytomel sodium, triiodothyronine sodium | |
| CHEBI:6484 | |
| C1=CC(=C(C=C1OC2=C(C=C(C=C2I)CC(C(=O)[O-])N)I)I)O.[Na+] |
Safety and Handling
EINECSNumber : 200-223-5
RTECSNumber : YP2836500
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only