missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole 98.0+%, TCI America™
$158.44 - $477.19
Chemical Identifiers
| CAS | 1345021-52-4 |
|---|---|
| Molecular Formula | C24H15Br2N |
| Molecular Weight (g/mol) | 477.20 |
| MDL Number | MFCD22571691 |
| InChI Key | GXVXADLFVMCWNC-UHFFFAOYSA-N |
| PubChem CID | 90027880 |
| IUPAC Name | 3-(3,5-dibromophenyl)-9-phenyl-9H-carbazole |
| SMILES | BrC1=CC(=CC(Br)=C1)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4979200MG
|
TCI America
D4979200MG |
200 mg |
Each for $158.44
|
|
|||||
|
D49791G
|
TCI America
D49791G |
1 g |
Each for $477.19
|
|
|||||
Chemical Identifiers
| 1345021-52-4 | |
| 477.20 | |
| GXVXADLFVMCWNC-UHFFFAOYSA-N | |
| 3-(3,5-dibromophenyl)-9-phenyl-9H-carbazole |
| C24H15Br2N | |
| MFCD22571691 | |
| 90027880 | |
| BrC1=CC(=CC(Br)=C1)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 |
Specifications
| 1345021-52-4 | |
| C24H15Br2N | |
| 200 mg | |
| BrC1=CC(=CC(Br)=C1)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 | |
| 477.20 | |
| 477.20 | |
| Crystalline Powder |
| 169°C | |
| MFCD22571691 | |
| GXVXADLFVMCWNC-UHFFFAOYSA-N | |
| 3-(3,5-dibromophenyl)-9-phenyl-9H-carbazole | |
| 90027880 | |
| ≥98.0% (GC) | |
| 3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole |
Safety and Handling
TSCA : No