missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole 98.0+%, TCI America™
Supplier: TCI America D4979200MG
Specifications
| 3-(3,5-Dibromophenyl)-9-phenyl-9H-carbazole | |
| 169°C | |
| MFCD22571691 | |
| GXVXADLFVMCWNC-UHFFFAOYSA-N | |
| 3-(3,5-dibromophenyl)-9-phenyl-9H-carbazole | |
| 90027880 | |
| ≥98.0% (GC) |
| 1345021-52-4 | |
| C24H15Br2N | |
| 200 mg | |
| BrC1=CC(=CC(Br)=C1)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 | |
| 477.20 | |
| 477.20 | |
| Crystalline Powder |
Chemical Identifiers
| 1345021-52-4 | |
| 477.20 | |
| GXVXADLFVMCWNC-UHFFFAOYSA-N | |
| 3-(3,5-dibromophenyl)-9-phenyl-9H-carbazole |
| C24H15Br2N | |
| MFCD22571691 | |
| 90027880 | |
| BrC1=CC(=CC(Br)=C1)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 |
Safety and Handling
TSCA : No