missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbiphenyl 98.0+%, TCI America™

Catalog No. p-7137494
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29481G 1 g
T29485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29481G Supplier TCI America Supplier No. T29481G
Only null left

Chemical Identifiers

CAS 25570-02-9
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00051910
InChI Key CMZYGFLOKOQMKF-UHFFFAOYSA-N
PubChem CID 520212
IUPAC Name 3,3',5,5'-tetramethyl-1,1'-biphenyl
SMILES CC1=CC(=CC(C)=C1)C1=CC(C)=CC(C)=C1

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Synonym 3,3',5,5'-tetramethylbiphenyl, 1-3,5-dimethylphenyl-3,5-dimethylbenzene, 3,5,3',5'-tetramethylbiphenyl, 1,1'-biphenyl, 3,3',5,5'-tetramethyl, biphenyl, 3,3',5,5'-tetramethyl, acmc-1cihy, 2,2'5.5'-tetramethylbiphenyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl,3,3',5,5'-tetramethyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl #
Formula Weight 210.32
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3,3′,5,5′-Tetramethylbiphenyl