missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',5,5'-Tetramethylbiphenyl, 97+%
Description
3,3',5,5'-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3',5,5'-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3',5,5'-tetracarboxylic acid by the oxidation of with potassium permanganate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,3′,5,5′-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3′,5,5′-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3′,5,5′-tetracarboxylic acid by the oxidation of with potassium permanganate.
Notes
Incompatible with strong oxidizing agents.
3,3′,5,5′-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3′,5,5′-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3′,5,5′-tetracarboxylic acid by the oxidation of with potassium permanganate.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 47°C to 49°C |
| Quantity | 1 g |
| Beilstein | 1935931 |
| Synonym | 3,3',5,5'-tetramethylbiphenyl, 1-3,5-dimethylphenyl-3,5-dimethylbenzene, 3,5,3',5'-tetramethylbiphenyl, 1,1'-biphenyl, 3,3',5,5'-tetramethyl, biphenyl, 3,3',5,5'-tetramethyl, acmc-1cihy, 2,2'5.5'-tetramethylbiphenyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl,3,3',5,5'-tetramethyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl # |
| Formula Weight | 210.32 |
| Percent Purity | ≥97% |
| Chemical Name or Material | 3,3',5,5'-Tetramethylbiphenyl |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only