missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbiphenyl, 97+%

Catalog No. AAL0917206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0917206 5 g
AAL0917203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0917206 Supplier Thermo Scientific Chemicals Supplier No. L0917206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25570-02-9 | C16H18 | 210.32 g/mol

3,3',5,5'-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3',5,5'-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3',5,5'-tetracarboxylic acid by the oxidation of with potassium permanganate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3′,5,5′-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3′,5,5′-tetracarboxylic acid by the oxidation of with potassium permanganate.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 25570-02-9
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00051910
InChI Key CMZYGFLOKOQMKF-UHFFFAOYSA-N
PubChem CID 520212
IUPAC Name 1-(3,5-dimethylphenyl)-3,5-dimethylbenzene
SMILES CC1=CC(=CC(C)=C1)C1=CC(C)=CC(C)=C1

Specifications

Melting Point 47°C to 49°C
Quantity 5 g
Beilstein 1935931
Synonym 3,3',5,5'-tetramethylbiphenyl, 1-3,5-dimethylphenyl-3,5-dimethylbenzene, 3,5,3',5'-tetramethylbiphenyl, 1,1'-biphenyl, 3,3',5,5'-tetramethyl, biphenyl, 3,3',5,5'-tetramethyl, acmc-1cihy, 2,2'5.5'-tetramethylbiphenyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl,3,3',5,5'-tetramethyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl #
Formula Weight 210.32
Percent Purity ≥97%
Chemical Name or Material 3,3',5,5'-Tetramethylbiphenyl
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only