missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbiphenyl-4,4'-diol 98.0+%, TCI America™

Catalog No. p-7137603
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31455G 5 g
T314525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31455G Supplier TCI America Supplier No. T31455G
Only null left

Chemical Identifiers

CAS 2417-04-1
Molecular Formula C16H18O2
Molecular Weight (g/mol) 242.32
MDL Number MFCD00094737
InChI Key YGYPMFPGZQPETF-UHFFFAOYSA-N
PubChem CID 75490
IUPAC Name 4-(4-hydroxy-3,5-dimethylphenyl)-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)C2=CC(=C(C(=C2)C)O)C

Specifications

Melting Point 225°C
Color Yellow
Quantity 5 g
Synonym 4,4′C-Dihydroxy-3,3′C,5,5′C-tetramethylbiphenyl
Formula Weight 242.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,5,5′-Tetramethylbiphenyl-4,4′-diol
EINECSNumber (4)-1675
TSCA Yes