Learn More
3,3',5,5'-Tetramethylbenzidine dihydrochloride, 98%
CAS: 207738-08-7 | C16H22Cl2N2 | 313.27 g/mol
Supplier: Thermo Scientific Chemicals J6033288
Description
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.
Solubility
Soluble in DMSO (∼1.7 mg/ml), and 1:300 DMSO:PBS (pH 7.2) (0.03 mg/ml).
Notes
Moisture and light sensitive. Store away from oxidizing agents, heat, light, acids, water/ moisture.
Specifications
| 3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate | |
| White | |
| MFCD00150104 | |
| Light and moisture sensitive | |
| Soluble in DMSO (∽1.7mg/ml),and 1:300 DMSO:PBS (pH 7.2) (0.03mg/ml). | |
| Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1 | |
| 313.27 | |
| 313.27 | |
| Powder |
| 207738-08-7 | |
| C16H22Cl2N2 | |
| 2.5 g | |
| 3,3',5,5'-tetramethylbenzidine dihydrochloride dihydrate, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride dihydrate, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, pubchem9044, tetramethylbenzidine dihydrate dihydrochloride | |
| NYNRGZULARUZCC-UHFFFAOYSA-N | |
| 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrate;dihydrochloride | |
| 19836601 | |
| ≥99% |
Chemical Identifiers
| 207738-08-7 | |
| 313.27 | |
| NYNRGZULARUZCC-UHFFFAOYSA-N | |
| 19836601 | |
| Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1 |
| C16H22Cl2N2 | |
| MFCD00150104 | |
| 3,3',5,5'-tetramethylbenzidine dihydrochloride dihydrate, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride dihydrate, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, pubchem9044, tetramethylbenzidine dihydrate dihydrochloride | |
| 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrate;dihydrochloride |
Safety and Handling
GHS H Statement
H341-H302-H315-H319
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H341
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only