missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, 98%

Catalog No. p-7081266
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1116403 1 g
AAA1116406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1116403 Supplier Thermo Scientific Chemicals Supplier No. A1116403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 175136-66-0 | C17H6F12O | 454.215 g/mol

3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, is used as reactive dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetrakis(trifluoromethyl)benzophenone, is used as reactive dye intermediates.

Solubility
Sparingly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 175136-66-0
Molecular Formula C17H6F12O
Molecular Weight (g/mol) 454.215
MDL Number MFCD00042474
InChI Key GATWMPGNBWPCIY-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetrakis trifluoromethyl benzophenone, bis 3,5-bis trifluoromethyl phenyl methanone, maybridge1_002937, acmc-1c7yy, bis 3,5-bis trifluoromethyl phenyl ketone, di 3,5-di trifluoromethyl phenyl methanone, bis 3,5-bis trifluoromethyl phenyl methanone #, 3,3/',5,5/'-tetrakis trifluoromethyl benzophenone
PubChem CID 519439
IUPAC Name bis[3,5-bis(trifluoromethyl)phenyl]methanone
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 139°C to 140°C
Quantity 1 g
Solubility Information Sparingly soluble in water.
Formula Weight 454.22
Percent Purity 98%
Chemical Name or Material 3,3',5,5'-Tetrakis (trifluoromethyl)benzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only