missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetrabromo-2,2'-bithiophene 98.0+%, TCI America™

Catalog No. T26995G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26995G 5 g
T26991G 1 g
T269925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T26995G Supplier TCI America Supplier No. T26995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 125143-53-5
Molecular Formula C8H2Br4S2
Molecular Weight (g/mol) 481.84
InChI Key MOMHMPZSZNZLAK-UHFFFAOYSA-N
PubChem CID 2740586
IUPAC Name 3,5-dibromo-2-(3,5-dibromothiophen-2-yl)thiophene
SMILES C1=C(SC(=C1Br)C2=C(C=C(S2)Br)Br)Br

Specifications

Melting Point 140°C
Quantity 5 g
Formula Weight 481.84
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,5,5′-Tetrabromo-2,2′-bithiophene