accessibility menu, dialog, popup

UserName

CAS RN 125143-53-5

IUPAC Name: 3,3',5,5'-tetrabromo-2,2'-bithiophene
Synonyms: 3,3',5,5'-tetrabromo-2,2'-bithiophene
Molecular Weight (g/mol): 481.84
Molecular Formula: C8H2Br4S2
InChi Key: MOMHMPZSZNZLAK-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C(S1)C1=C(Br)C=C(Br)S1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results