missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dichlorophenyl)-1,1-dimethylurea 98.0+%, TCI America™

Supplier:  TCI America D1328500G

 View more versions of this product

Catalog No. D1328500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-(3,4-Dichlorophenyl)-1,1-dimethylurea
159°C
C9H10Cl2N2O
500 g
diuron, 3-3,4-dichlorophenyl-1,1-dimethylurea, dcmu, dynex, duran, dichlorfenidim, herbatox, vonduron, dailon, karmex
CN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl
233.092
CHEBI:116509
≥98.0% (N)
330-54-1
White-Yellow
MFCD00018136
3077
XMTQQYYKAHVGBJ-UHFFFAOYSA-N
3-(3,4-dichlorophenyl)-1,1-dimethylurea
3120
233.09
Crystalline Powder

Chemical Identifiers

330-54-1
233.092
XMTQQYYKAHVGBJ-UHFFFAOYSA-N
3120
3-(3,4-dichlorophenyl)-1,1-dimethylurea
C9H10Cl2N2O
MFCD00018136
diuron, 3-3,4-dichlorophenyl-1,1-dimethylurea, dcmu, dynex, duran, dichlorfenidim, herbatox, vonduron, dailon, karmex
CHEBI:116509
CN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2194

RTECSNumber : YS8925000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions