missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dichlorophenyl)-1,1-dimethylurea 98.0+%, TCI America™

Catalog No. D132825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D132825G 25 g
D1328500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D132825G Supplier TCI America Supplier No. D132825G

Chemical Identifiers

CAS 330-54-1
Molecular Formula C9H10Cl2N2O
Molecular Weight (g/mol) 233.092
MDL Number MFCD00018136
InChI Key XMTQQYYKAHVGBJ-UHFFFAOYSA-N
Synonym diuron, 3-3,4-dichlorophenyl-1,1-dimethylurea, dcmu, dynex, duran, dichlorfenidim, herbatox, vonduron, dailon, karmex
PubChem CID 3120
ChEBI CHEBI:116509
IUPAC Name 3-(3,4-dichlorophenyl)-1,1-dimethylurea
SMILES CN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 233.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3-(3,4-Dichlorophenyl)-1,1-dimethylurea
EINECSNumber (3)-2194
RTECSNumber YS8925000
TSCA Yes