missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dichlorophenyl)-1,1-dimethylurea, 97%

Catalog No. p-7043834
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0298618 50 g
AAL0298630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL0298618 Supplier Thermo Scientific Chemicals Supplier No. L0298618
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 330-54-1 | C9H10Cl2N2O | 233.092 g/mol

A phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff

Solubility
Soluble in water (0.049g/L), chloroform and methanol. Very slightly soluble in many organic solvents.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 330-54-1
Molecular Formula C9H10Cl2N2O
Molecular Weight (g/mol) 233.092
MDL Number MFCD00018136
InChI Key XMTQQYYKAHVGBJ-UHFFFAOYSA-N
Synonym diuron, 3-3,4-dichlorophenyl-1,1-dimethylurea, dcmu, dynex, duran, dichlorfenidim, herbatox, vonduron, dailon, karmex
PubChem CID 3120
ChEBI CHEBI:116509
IUPAC Name 3-(3,4-dichlorophenyl)-1,1-dimethylurea
SMILES CN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl

Specifications

Melting Point 157°C to 160°C
Density 1.48
Boiling Point 180°C to 190°C (decomposition)
Odor Odorless
Quantity 50 g
UN Number UN3077
Beilstein 2215168
Merck Index 14,3382
Solubility Information Soluble in water (0.049g/L),chloroform and methanol. Very slightly soluble in many organic solvents.
Formula Weight 233.1
Percent Purity 97%
Chemical Name or Material 3-(3,4-Dichlorophenyl)-1,1-dimethylurea
Show More Show Less
Hazard Category H302-H351-H373
Hazard Statement GHS H Statement
H351-H373-H302
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 206-354-4
RTECSNumber YS8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only