missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4'-Tetramethylbiphenyl, 98%

Catalog No. AAL1333803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1333803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1333803 Supplier Thermo Scientific Chemicals Supplier No. L1333803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4920-95-0 | C16H18 | 210.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4920-95-0
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00130219
InChI Key YXBIAYXZUDJVEB-UHFFFAOYSA-N
Synonym 3,3',4,4'-tetramethylbiphenyl, 3,3',4,4'-tetramethyl-1,1'-biphenyl, 3,4,3',4'-tetramethylbiphenyl, 1,1'-biphenyl, 3,3',4,4'-tetramethyl, biphenyl, 3,3',4,4'-tetramethyl, 3,3',4,4'-tetramethyldiphenyl, 4-3,4-dimethylphenyl-1,2-dimethylbenzene, acmc-20anh7, amtda180, 4-05-00-01956 beilstein handbook reference
PubChem CID 21029
IUPAC Name 4-(3,4-dimethylphenyl)-1,2-dimethylbenzene
SMILES CC1=CC=C(C=C1C)C1=CC=C(C)C(C)=C1

Specifications

Melting Point 74°C to 77°C
Quantity 1 g
Beilstein 1935370
Formula Weight 210.32
Percent Purity 98%
Chemical Name or Material 3,3',4,4'-Tetramethylbiphenyl
RTECSNumber DV8730000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only