missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4'-Benzophenonetetracarboxylic Dianhydride 96.0+%, TCI America™

Catalog No. B094825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B094825G 25 g
B0948100G 100 g
B0948500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B094825G Supplier TCI America Supplier No. B094825G

Chemical Identifiers

CAS 2421-28-5
Molecular Formula C17H6O7
Molecular Weight (g/mol) 322.228
MDL Number MFCD00005923
InChI Key VQVIHDPBMFABCQ-UHFFFAOYSA-N
Synonym 3,3',4,4'-benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic acid dianhydride, 1,3-isobenzofurandione, 5,5'-carbonylbis, 4,4'-carbonylbis phthalic anhydride, 4,4'-carbonyldiphthalic anhydride, 4,4'-diphthalic anhydride ketone, benzophenonetetracarboxylic anhydride, phthalic anhydride, 4,4'-carbonyldi, unii-y61gva8097
PubChem CID 75498
IUPAC Name 5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 25 g
Formula Weight 322.23
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,4,4′-Benzophenonetetracarboxylic Dianhydride
EINECSNumber (4)-0150
TSCA Yes