missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 401930025

Quantity 2.5kg
Packaging Plastic Bottle

 View more versions of this product

Catalog No. AC401930025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2421-28-5
322.22
VQVIHDPBMFABCQ-UHFFFAOYSA-N
75498
C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
C17H6O7
MFCD00005923
3,3',4,4'-benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic acid dianhydride, 1,3-isobenzofurandione, 5,5'-carbonylbis, 4,4'-carbonylbis phthalic anhydride, 4,4'-carbonyldiphthalic anhydride, 4,4'-diphthalic anhydride ketone, benzophenonetetracarboxylic anhydride, phthalic anhydride, 4,4'-carbonyldi, unii-y61gva8097
5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione

Specifications

3,3′,4,4′-Benzophenonetetracarboxylic dianhydride
2421-28-5
100.0
97+%
C17H6O7
MFCD00005923
0220.0°C
VQVIHDPBMFABCQ-UHFFFAOYSA-N
5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
75498
97+%
99%
98.5
Authentic
Plastic Bottle
2.5kg
3,3',4,4'-benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic acid dianhydride, 1,3-isobenzofurandione, 5,5'-carbonylbis, 4,4'-carbonylbis phthalic anhydride, 4,4'-carbonyldiphthalic anhydride, 4,4'-diphthalic anhydride ketone, benzophenonetetracarboxylic anhydride, phthalic anhydride, 4,4'-carbonyldi, unii-y61gva8097
Solubility in water: reacts. Other solubilities: soluble in dmf (1g/10mL)
C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
322.22
322.22
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

EINECSNumber : 219-348-1

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...