missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific™

Catalog No. p-63687
Change view
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401935000 500g Glass bottle
AC401930025 2.5kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC401935000 Supplier Thermo Scientific Chemicals Supplier No. 401935000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 2421-28-5
Molecular Formula C17H6O7
Molecular Weight (g/mol) 322.22
MDL Number MFCD00005923
InChI Key VQVIHDPBMFABCQ-UHFFFAOYSA-N
Synonym 3,3',4,4'-benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic acid dianhydride, 1,3-isobenzofurandione, 5,5'-carbonylbis, 4,4'-carbonylbis phthalic anhydride, 4,4'-carbonyldiphthalic anhydride, 4,4'-diphthalic anhydride ketone, benzophenonetetracarboxylic anhydride, phthalic anhydride, 4,4'-carbonyldi, unii-y61gva8097
PubChem CID 75498
IUPAC Name 5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 500g
Sublimation Point 0220.0°C
Solubility Information Solubility in water: reacts. Other solubilities: soluble in dmf (1g/10mL)
Formula Weight 322.22
Percent Purity 97+%
Chemical Name or Material 3,3′,4,4′-Benzophenonetetracarboxylic dianhydride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
EINECSNumber 219-348-1
TSCA TSCA

RUO – Research Use Only