missing translation for 'onlineSavingsMsg'
Learn More

3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexene 98.0+%, TCI America™

Catalog No. p-7133887
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06015G 5 g
N060125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06015G Supplier TCI America Supplier No. N06015G
Only null left

Chemical Identifiers

CAS 19430-93-4
Molecular Formula C6H3F9
Molecular Weight (g/mol) 246.076
MDL Number MFCD00042338
InChI Key GVEUEBXMTMZVSD-UHFFFAOYSA-N
Synonym perfluorobutyl ethylene, 3,3,4,4,5,5,6,6,6-nonafluoro-1-hexene, perfluorobutylethylene, 1h,1h,2h-perfluoro-1-hexene, 1h,1h,2h-perfluorohexene, perfluorobutyl ethene, 1-hexene, 3,3,4,4,5,5,6,6,6-nonafluoro, 3,3,4,4,5,5,6,6,6-nonafluorohexene, perfluoro-n-butyl ethylene, 1h,1h,2h-perfluorohex-1-ene
PubChem CID 88054
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohex-1-ene
SMILES C=CC(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Red
Boiling Point 60°C
Quantity 5 g
UN Number 1993
Formula Weight 246.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexene
EINECSNumber (2)-3339
RTECSNumber MP7360000
TSCA Yes