accessibility menu, dialog, popup

UserName

CAS RN 19430-93-4

IUPAC Name: 3,3,4,4,5,5,6,6,6-nonafluorohex-1-ene
Synonyms: 3,3,4,4,5,5,6,6,6-nonafluorohex-1-ene
Molecular Weight (g/mol): 246.08
Molecular Formula: C6H3F9
InChi Key: GVEUEBXMTMZVSD-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)