missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Triphenylpropionic Acid 98.0+%, TCI America™

Catalog No. T15585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15585G 5 g
T155825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15585G Supplier TCI America Supplier No. T15585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 900-91-4
Molecular Formula C21H18O2
Molecular Weight (g/mol) 302.373
MDL Number MFCD00002713
InChI Key XMSJLUKCGWQAHO-UHFFFAOYSA-N
Synonym 3,3,3-triphenylpropionic acid, tritylacetic acid, 3,3,3-triphenyl propanoic acid, 2-tritylacetic acid, pubchem12502, acmc-209r4v, 2-triphenylmethylacetic acid, 2-triphenylmethyl acetic acid, 3,3 3-triphenyl propionic acid
PubChem CID 136696
IUPAC Name 3,3,3-triphenylpropanoic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 181°C
Quantity 5 g
Formula Weight 302.37
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3,3-Triphenylpropionic Acid