missing translation for 'onlineSavingsMsg'
Learn More

(3,3,3-Trifluoropropyl)trimethoxysilane, 97%

Catalog No. AAL1667114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1667114 25 g
AAL1667106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1667114 Supplier Thermo Scientific Chemicals Supplier No. L1667114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 429-60-7 | C6H13F3O3Si | 218.25 g/mol

(3,3,3-Trifluoropropyl)trimethoxysilane is used in the manufacture of defoaming agents, releasing agents, fluoro silicone resin and coating. It is used as a coupling agent for epoxy resin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3,3,3-Trifluoropropyl)trimethoxysilane is used in the manufacture of defoaming agents, releasing agents, fluoro silicone resin and coating. It is used as a coupling agent for epoxy resin.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.

Chemical Identifiers

CAS 429-60-7
Molecular Formula C6H13F3O3Si
Molecular Weight (g/mol) 218.25
MDL Number MFCD00039266
InChI Key JLGNHOJUQFHYEZ-UHFFFAOYSA-N
Synonym trimethoxy 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trimethoxysilane, 3,3,3-trifluoropropyltrimethoxysilane, silane, trimethoxy 3,3,3-trifluoropropyl, 1,1,1-trifluoro-3-trimethoxysilyl propane, silane, trimethoxy trifluoropropyl, silane,trimethoxy 3,3,3-trifluoropropyl, acmc-1ajbz, fas-3, cf3 ch2 2si och3 3
PubChem CID 67933
IUPAC Name trimethoxy(3,3,3-trifluoropropyl)silane
SMILES CO[Si](CCC(F)(F)F)(OC)OC

Specifications

Boiling Point 144°C
Quantity 25 g
UN Number UN1993
Beilstein 1762962
Flash Point 38°C (100°F)
Formula Weight 218.25
Percent Purity 97%
Refractive Index 1.355
Sensitivity Moisture sensitive
Density 1.14
Chemical Name or Material (3,3,3-Trifluoropropyl)trimethoxysilane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 207-059-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only