missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%

Catalog No. p-3779783
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397860010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC397860010 Supplier Thermo Scientific Chemicals Supplier No. 397860010

CAS: 564-10-3 | C4H2F6O2 | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 564-10-3
Molecular Formula C4H2F6O2
Molecular Weight (g/mol) 196.05
MDL Number MFCD00464165
InChI Key RAEAYTICAPHWJW-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid
PubChem CID 385606
IUPAC Name 3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
SMILES C(C(=O)O)(C(F)(F)F)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50°C to 53°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CF3)2CHCOOH
Quantity 1 g
Formula Weight 196.05
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3, 3-Trifluoro-2-(trifluoromethyl)propionic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

RUO – Research Use Only