accessibility menu, dialog, popup

UserName

CAS RN 564-10-3

IUPAC Name: 3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
Synonyms: 3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
Molecular Weight (g/mol): 196.05
Molecular Formula: C4H2F6O2
InChi Key: RAEAYTICAPHWJW-UHFFFAOYSA-N
SMILES: OC(=O)C(C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)