missing translation for 'onlineSavingsMsg'
Learn More

3,3',3″-Nitrilotris(propionamide) 98.0+%, TCI America™

Catalog No. N0342500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0342500G 500 g
N034225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0342500G Supplier TCI America Supplier No. N0342500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2664-61-1
Molecular Formula C9H18N4O3
Molecular Weight (g/mol) 230.268
MDL Number MFCD00059194
InChI Key RERXJGPPGMABOY-UHFFFAOYSA-N
PubChem CID 75874
IUPAC Name 3-[bis(3-amino-3-oxopropyl)amino]propanamide
SMILES C(CN(CCC(=O)N)CCC(=O)N)C(=O)N

Specifications

Melting Point 183°C
Color White
Quantity 500 g
Formula Weight 230.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,3′′-Nitrilotris(propionamide)
EINECSNumber (2)-1278
TSCA Yes