accessibility menu, dialog, popup

UserName

NH2ONNH2ONH2O

CAS RN 2664-61-1

IUPAC Name: 3-[bis(2-carbamoylethyl)amino]propanamide
Synonyms: 3-[bis(2-carbamoylethyl)amino]propanamide
Molecular Weight (g/mol): 230.27
Molecular Formula: C9H18N4O3
InChi Key: RERXJGPPGMABOY-UHFFFAOYSA-N
SMILES: NC(=O)CCN(CCC(N)=O)CCC(N)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)